Learn More
Cell Signaling Technology MEK1/2 Control Cell Extracts, Western Blotting
SDP
Catalog No. 50195189
Encompass
These MEK1/2 Control Cell Extracts provide essential positive and negative control lysates for western blotting applications. The set includes nonphosphorylated total cell extracts derived from serum-starved HeLa cells as a negative control, and phosphorylated total cell extracts from HeLa cells treated with TPA to serve as a positive control. The extracts are supplied in SDS sample buffer for immediate use.
  • Includes both positive and negative controls for reliable data validation
  • Suitable for western blotting applications
  • Supplied in ready-to-use SDS sample buffer
  • Validated cell extract format from HeLa cell lines
Catalog No. 50-195-189 Supplier Cell Signaling Technology Supplier No. 9160S
These MEK1/2 Control Cell Extracts provide essential positive and negative control lysates for western blotting applications. The set includes nonphosphorylated total cell extracts derived from serum-starved HeLa cells as a negative control, and phosphorylated total cell extracts from HeLa cells treated with TPA to serve as a positive control. The extracts are supplied in SDS sample buffer for immediate use.
  • Includes both positive and negative controls for reliable data validation
  • Suitable for western blotting applications
  • Supplied in ready-to-use SDS sample buffer
  • Validated cell extract format from HeLa cell lines