Learn More
Abcam Cell Fractionation Kit - High Throughput (HT), 1 x 96 TestsS
Catalog No. NC1988810
Encompass_Preferred
Abcam's Cell Fractionation Kit - High Throughput (HT), provides a method and reagents for a simple and rapid preparation of cytosolic, mitochondrial and nuclear fractions from adherent cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1988810 Supplier Abcam Supplier No. AB1097181X96TESTS
Abcam's Cell Fractionation Kit - High Throughput (HT), provides a method and reagents for a simple and rapid preparation of cytosolic, mitochondrial and nuclear fractions from adherent cells.
The product is subject to the following: Abcam Restricted Use Statement