Learn More
Abcam Cell Fractionation Kit - Standard, 1KIT
Catalog No. NC1241258
Encompass_Preferred
Cell fractionation kit (standard) allows for the rapid and simple preparation of mitochondrial, cytoplasmic, and nuclear fractions from cultured cells of mammals and other species. - Cited in over 120 publications - Quickly isolates mitochondrial, cytoplasmic, and nuclear fractions without the need for mechanical disruption - Provides highly enriched fractions for monitoring proteins of interest across different cellular compartments - Useful for studying apoptosis and pro-apoptotic protein movement
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1241258 Supplier Abcam Supplier No. AB1097191KIT
Cell fractionation kit (standard) allows for the rapid and simple preparation of mitochondrial, cytoplasmic, and nuclear fractions from cultured cells of mammals and other species. - Cited in over 120 publications - Quickly isolates mitochondrial, cytoplasmic, and nuclear fractions without the need for mechanical disruption - Provides highly enriched fractions for monitoring proteins of interest across different cellular compartments - Useful for studying apoptosis and pro-apoptotic protein movement
The product is subject to the following: Abcam Restricted Use Statement