Learn More
Biochain Institute Inc cfPure LG Cell Free DNA Extraction Kit (100 ml)
SDP
Catalog No. 501871531
Encompass_Preferred
BioChain's cfPure Cell Free DNA Extraction Kit is designed for rapid & efficient purification of circulating cell free DNA (cfDNA). The system utilizes silica-coated paramagnetic particles to purify cell free DNA from less than 1 mL to greater than 10 mL of serum or plasma. The buffers were developed to ensure efficient recovery of 50 bp - 500 bp DNA fragments in order to maximize recovery of cell free DNA. The recovered DNA is suitable for a wide range of down-stream applications, such as bisulfite sequencing, NGS, & qPCR. The cfPure protocol is rapid & easy, allowing users to process several samples in an hour or less & making it ideal for biomarker screening. BioChain has developed its own silica-coated magnetic bead technology, which is designed to efficiently recover low molecular weight cell free DNA during isolation. This kit is automation friendly and there are several versions for the kit format. LG is for "dirty" plasma in order to eliminate genomic DNA contamination.
Catalog No. 50-187-1531 Supplier Biochain Institute Inc Supplier No. K5011610V2
BioChain's cfPure Cell Free DNA Extraction Kit is designed for rapid & efficient purification of circulating cell free DNA (cfDNA). The system utilizes silica-coated paramagnetic particles to purify cell free DNA from less than 1 mL to greater than 10 mL of serum or plasma. The buffers were developed to ensure efficient recovery of 50 bp - 500 bp DNA fragments in order to maximize recovery of cell free DNA. The recovered DNA is suitable for a wide range of down-stream applications, such as bisulfite sequencing, NGS, & qPCR. The cfPure protocol is rapid & easy, allowing users to process several samples in an hour or less & making it ideal for biomarker screening. BioChain has developed its own silica-coated magnetic bead technology, which is designed to efficiently recover low molecular weight cell free DNA during isolation. This kit is automation friendly and there are several versions for the kit format. LG is for "dirty" plasma in order to eliminate genomic DNA contamination.