Learn More
Agilent Technologies Cell-free DNA Reagents, Assay Type: cfDNA, DNA Analysis Range: 50 to 800 bp, Sample Capacity: 112 samples, Regulatory Status: Research Use Only (RUO), Guaranteed Shelf Life: 6 months
Catalog No. NC2380479
Encompass
The Agilent Technologies Cell-free DNA Reagents are designed for precise analysis of cell-free DNA (cfDNA) utilizing Agilent TapeStation systems. These reagents allow for integrity assessment, quantification, and sizing of cfDNA samples in the 50 to 800 bp range, including detection of high molecular weight DNA contamination. Intended for research use only, the product offers an efficient and automated solution for nucleic acid quality control workflows, particularly in next-generation sequencing (NGS) applications.
  • Automated and efficient analysis minimizes hands-on time.
  • Enables comprehensive cfDNA characterization, including quantification and sizing.
  • High sensitivity detection down to 20 pg/μL ensures accurate analysis.
  • Streamlined workflow compatible with Agilent TapeStation systems (4150, 4200).
  • Supports a sample capacity of 112 samples.
  • Guaranteed shelf life is 6 months.
Catalog No. NC2380479 Supplier Agilent Technologies Supplier No. 50675631
The Agilent Technologies Cell-free DNA Reagents are designed for precise analysis of cell-free DNA (cfDNA) utilizing Agilent TapeStation systems. These reagents allow for integrity assessment, quantification, and sizing of cfDNA samples in the 50 to 800 bp range, including detection of high molecular weight DNA contamination. Intended for research use only, the product offers an efficient and automated solution for nucleic acid quality control workflows, particularly in next-generation sequencing (NGS) applications.
  • Automated and efficient analysis minimizes hands-on time.
  • Enables comprehensive cfDNA characterization, including quantification and sizing.
  • High sensitivity detection down to 20 pg/μL ensures accurate analysis.
  • Streamlined workflow compatible with Agilent TapeStation systems (4150, 4200).
  • Supports a sample capacity of 112 samples.
  • Guaranteed shelf life is 6 months.