Learn More
Sino Biological Cell-Free Protein Synthesis kit 50 Tests
Catalog No. 5000696243
Encompass_Preferred
CFKIT02 is a cell-free protein synthesis system based on E. coli lysate, supplemented with all the enzymes, energy regeneration system, and buffer environment required for efficient in vitro transcription and translation. Users only need to provide plasmid or PCR product templates to obtain soluble proteins within hours.
Catalog No. 50-006-96243 Supplier Sino Biological Supplier No. CFKIT0250
CFKIT02 is a cell-free protein synthesis system based on E. coli lysate, supplemented with all the enzymes, energy regeneration system, and buffer environment required for efficient in vitro transcription and translation. Users only need to provide plasmid or PCR product templates to obtain soluble proteins within hours.