Learn More
Sciencell Research Laboratories CELL FREEZING MEDIUM 50ML
Catalog No. NC9664283
Encompass
CFM is a sterile, uniquely formulated cell freezing medium that significantly improves cell viability and function following cryopreservation. It consists of 10% fetal bovine serum, 10% DMSO and 10 mM HEPES in modified MCDB 131 medium.
Catalog No. NC9664283 Supplier Sciencell Research Laboratories Supplier No. 0133
CFM is a sterile, uniquely formulated cell freezing medium that significantly improves cell viability and function following cryopreservation. It consists of 10% fetal bovine serum, 10% DMSO and 10 mM HEPES in modified MCDB 131 medium.