Learn More
Quality Biological Inc Cell Freezing Medium B 2X (DMSO Freezing Medium), 4 x 50ML
SDP
Catalog No. 50983224
Encompass_Preferred
Cell freezing and storage at sub-zero temperatures is an important method for preserving cells for future studies. Cell freezing or cryopreservation ensures that you have a back-up of cells in case of contamination and loss of cell supply. It is best to cryopreserve cells when they are at their maximum growth rate or almost confluent. Cell Freezing Medium B when used with Cell Freezing Medium A protects cells from damage after freezing and thawing.
Catalog No. 50-983-224 Supplier Quality Biological Inc Supplier No. 118257711
Add to Cart
Add to Cart
Cell freezing and storage at sub-zero temperatures is an important method for preserving cells for future studies. Cell freezing or cryopreservation ensures that you have a back-up of cells in case of contamination and loss of cell supply. It is best to cryopreserve cells when they are at their maximum growth rate or almost confluent. Cell Freezing Medium B when used with Cell Freezing Medium A protects cells from damage after freezing and thawing.