Learn More
Sigma Aldrich Fine Chemicals Biosciences Cell freezing Medium-DMSO
Catalog No. 502900349
Encompass
Cell freezing medium is a combination of cryoprotectant DMSO with fetal bovine serum. However serum free medium replaces serum with methylcellulose and poloxamer 188 and other additives like ascorbic acid tocopherol and proline.
Catalog No. 50-290-0349 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C629550ML
Cell freezing medium is a combination of cryoprotectant DMSO with fetal bovine serum. However serum free medium replaces serum with methylcellulose and poloxamer 188 and other additives like ascorbic acid tocopherol and proline.