Learn More
Perkin Elmer US LLC Cell Holder Cap
Catalog No. 502112325
Encompass_Preferred
The Quartz Cell with Low Profile Stopper is the only cell that can be used in the Integrating Fluorescence Spheres (N4201017). It's low profile avoids sample spills that could damage the Spheres.
Catalog No. 50-211-2325 Supplier Perkin Elmer US LLC Supplier No. N4202031
The Quartz Cell with Low Profile Stopper is the only cell that can be used in the Integrating Fluorescence Spheres (N4201017). It's low profile avoids sample spills that could damage the Spheres.