Learn More
Starna Cells Inc CELL HOLDER/CH2049
SDP
Catalog No. NC9594812
Encompass_Preferred
Cell Holder, Exterior Dimension: 12.5 x 12.5 x 52mm, Base Thickness: 9mm, For Use in a Standard Instrument
Catalog No. NC9594812 Supplier Starna Cells Inc Supplier No. CH2049
Cell Holder, Exterior Dimension: 12.5 x 12.5 x 52mm, Base Thickness: 9mm, For Use in a Standard Instrument