Learn More
Perkin Elmer US LLC Rectangular Long Path Cell Holder for LAMBDA Spectrometers
Catalog No. 509042126
Encompass_Preferred
This optional cell holder is easy to install and has the same footprint as the Standard Cell Holder. It is designed for a precise fit and gives you maximum reproducibility. Compatible with the LAMBDA 20/30/40/20Bio/30Bio/40Bio/ 650/750/850/900/950/1050/1050+. An additional holder can be ordered to accommodate the second beam on these spectrometers.
Catalog No. 50-904-2126 Supplier Perkin Elmer US LLC Supplier No. B0080821
This optional cell holder is easy to install and has the same footprint as the Standard Cell Holder. It is designed for a precise fit and gives you maximum reproducibility. Compatible with the LAMBDA 20/30/40/20Bio/30Bio/40Bio/ 650/750/850/900/950/1050/1050+. An additional holder can be ordered to accommodate the second beam on these spectrometers.