Learn More
Fluidigm Corp Cell-ID™ Intercalator-Rh, 2000 µM
SDP
Catalog No. NC1605684
Encompass
Cell-ID Intercalator-103Rh is a cationic nucleic acid intercalator that contains natural-abundance rhodium (103Rh) and is used in CyTOF analysis for discrimination of dead cells from live cells (if staining is done before cell fixation) or to discriminate single nucleated cells from doublets (if staining is done post-fixation). It is a live cell membrane-impermeable dye. For CyTOF2 and Helios™ instruments we recommend using Cell-ID Intercalator-Ir (201192B) for nucleated cell discrimination and Cisplatin-194 (201194) or -198 (201198) for dead cell discrimination.
Catalog No. NC1605684 Supplier Fluidigm Corp Supplier No. 201103B
Cell-ID Intercalator-103Rh is a cationic nucleic acid intercalator that contains natural-abundance rhodium (103Rh) and is used in CyTOF analysis for discrimination of dead cells from live cells (if staining is done before cell fixation) or to discriminate single nucleated cells from doublets (if staining is done post-fixation). It is a live cell membrane-impermeable dye. For CyTOF2 and Helios™ instruments we recommend using Cell-ID Intercalator-Ir (201192B) for nucleated cell discrimination and Cisplatin-194 (201194) or -198 (201198) for dead cell discrimination.