Learn More
Yamato Scientific America, Inc. Cell Imager BF + Red Fluo 10x
Catalog No. NC3812012 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Celloger Mini Plus is an automated live cell imaging system that is equipped with an advanced fluorescence and bright field microscopy, autofocusing and real time multi-position imaging technology. Allows easy monitoring of live cells inside the incubator for a long time without disturbing the environment suitable for cell culture. It doesn’t have a moveable stage but instead, the camera inside the system moves to capture the images of cell in multiple positions. Since the vessel and cell sample are in a steady state, this provides a stable environment for the cells to grow. Its compact size can fit into a standard CO2 incubator and can easily be carried in and out of the incubator during the experiment.
Catalog No. NC3812012 Supplier Yamato Scientific America, Inc. Supplier No. CMPBR10
Add to Cart
Add to Cart
Request a Quote
Celloger Mini Plus is an automated live cell imaging system that is equipped with an advanced fluorescence and bright field microscopy, autofocusing and real time multi-position imaging technology. Allows easy monitoring of live cells inside the incubator for a long time without disturbing the environment suitable for cell culture. It doesn’t have a moveable stage but instead, the camera inside the system moves to capture the images of cell in multiple positions. Since the vessel and cell sample are in a steady state, this provides a stable environment for the cells to grow. Its compact size can fit into a standard CO2 incubator and can easily be carried in and out of the incubator during the experiment.