Learn More
Spectrocell Inc CELL INJETABLE W/CASE REC SCRW
Catalog No. NC9781628
Encompass
Screw Top Cells, 10mm L, Volume: 3.5mL, PTFE Screw Taps, Injectable Cap, 2 Polished Windows, Spectral Range 170-2200nm
Catalog No. NC9781628 Supplier Spectrocell Inc Supplier No. R3010T
Screw Top Cells, 10mm L, Volume: 3.5mL, PTFE Screw Taps, Injectable Cap, 2 Polished Windows, Spectral Range 170-2200nm