Learn More
STEMCELL Technologies Cell Lifter with Flat Blade and J-Hook, Double End, Size: 100 Units
SDP
Catalog No. 502390174
Encompass
Use Cell Scraper Double End for a variety of cell culture applications. Made of high-density polyethylene (HDPE) each cell lifter comes individually wrapped and is sterile non-pyrogenic and RNase/DNase-free. The double end design features a flat blade on one end and either a narrow blade or a narrow J-hook blade on the other. The flat blade makes even contact with growth surfaces and is ideal for harvesting cells in Tissue Culture-Treated Dishes (Catalog 38046) and flat-bottom plates (e.g. 6-well Catalog 38015/38016). The narrow blade is optimized to fit 96-well plates (e.g. Catalog 38022) and can also be used for resuspending cell pellets in microcentrifuge tubes. The narrow J-hook blade can be used to vertically lift cells from 24-well plates (e.g. Catalog 38017/38021).
Catalog No. 50-239-0174 Supplier STEMCELL Technologies Supplier No. 2000597
Use Cell Scraper Double End for a variety of cell culture applications. Made of high-density polyethylene (HDPE) each cell lifter comes individually wrapped and is sterile non-pyrogenic and RNase/DNase-free. The double end design features a flat blade on one end and either a narrow blade or a narrow J-hook blade on the other. The flat blade makes even contact with growth surfaces and is ideal for harvesting cells in Tissue Culture-Treated Dishes (Catalog 38046) and flat-bottom plates (e.g. 6-well Catalog 38015/38016). The narrow blade is optimized to fit 96-well plates (e.g. Catalog 38022) and can also be used for resuspending cell pellets in microcentrifuge tubes. The narrow J-hook blade can be used to vertically lift cells from 24-well plates (e.g. Catalog 38017/38021).