Learn More
StatLab Cell Line Block ALK Lymphoma Analyte Control 2 Core
SDP
Catalog No. 21033317
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
21-033-317 1 Ea.
1 options
Catalog No. 21-033-317 Supplier StatLab Supplier No. CBLKNPMALK2
A quality control product used in laboratories for immunohistochemistry (IHC) and in situ hybridization (ISH) testing
  • 1 Cell Line Control Block
  • Type: ALK Lymphoma Analyte Control 2 Core
  • It consists of a cell microarray block with two cores: one that is positive for the NPM-ALK translocation and one that is negative, which is crucial for verifying that the testing reagents and protocols are working correctly for identifying ALK-positive lymphomas
Specifications
Autoclavable Not Autoclavable
Quantity 1 Ea.