Learn More
FUJIFILM Biosciences MKN1 cell line, JCRB0252, adherent growth, human gastric adenosquamous carcinoma, 72 years old male, stomach source, BSL 1, RPMI 1640 medium, 10% FBS, 37 °C, 5% CO2
Catalog No. NC2873599
Encompass
MKN1 cell line, JCRB0252, adherent growth, human gastric adenosquamous carcinoma, 72 years old male, stomach source, BSL 1, RPMI 1640 medium, 10% FBS, 37 °C, 5% CO2
Catalog No. NC2873599 Supplier FUJIFILM Biosciences Supplier No. JCRB0252
MKN1 cell line, JCRB0252, adherent growth, human gastric adenosquamous carcinoma, 72 years old male, stomach source, BSL 1, RPMI 1640 medium, 10% FBS, 37 °C, 5% CO2