Learn More
Lonza Walkersville Cell Line Nucleofector Kit L
Catalog No. NC9823333
Encompass
The Cell Line Nucleofector Kit L is one of our five Nucleofector Kits suited for transfecting cell lines in the Nucleofector 2b Device.This kit has been determined to be optimal for transfecting following cell types:3T3-L1 adA-10BHK-21BxPC-3DU145EL4FDC-P1GH3 H9c2 (2-1)HeLa S3IMR-32KG-1aL-428 (DSMZ)MDCKMDCK II (ECACC)MOLT-4MV-4-11NTERA-2 cl.D1WEHI-231The respective cell type-specific Optimized Protocol or recommendation is available under "Instructions" or in our knowledge database. In this Optimized Protocol the best Nucleofection conditions are indicated. In addition, we share our experience and knowledge for treatment of this cell type.Benefits:High transfection efficiencies in difficult-to-transfect cell linesApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells Low-throughput transfection in single cuvette format
Catalog No. NC9823333 Supplier Lonza Walkersville Supplier No. VCA1005
The Cell Line Nucleofector Kit L is one of our five Nucleofector Kits suited for transfecting cell lines in the Nucleofector 2b Device.This kit has been determined to be optimal for transfecting following cell types:3T3-L1 adA-10BHK-21BxPC-3DU145EL4FDC-P1GH3 H9c2 (2-1)HeLa S3IMR-32KG-1aL-428 (DSMZ)MDCKMDCK II (ECACC)MOLT-4MV-4-11NTERA-2 cl.D1WEHI-231The respective cell type-specific Optimized Protocol or recommendation is available under "Instructions" or in our knowledge database. In this Optimized Protocol the best Nucleofection conditions are indicated. In addition, we share our experience and knowledge for treatment of this cell type.Benefits:High transfection efficiencies in difficult-to-transfect cell linesApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells Low-throughput transfection in single cuvette format