Learn More
Thermo Scientific™ Cell Locker™ Chamber with Copper Tray, Stainless Steel
Catalog No. 11675973 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Places:
53
Catalog No. No. of Places
11-675-973 53
1 options
Catalog No. 11-675-973 Supplier Thermo Scientific™ Supplier No. 50154739
  • $1,814.00
    $1,548.65 / Each of 1
    Save $265.35
    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Protected chambers for your most sensitive cells
  • Each individual Cell Locker Chamber is effectively a quarantine chamber.
  • Dual 0.2μm membrane filters permits air circulation but exclude microbial contaminants.
  • The replacable 0.2μm membrane filter is hydrophobic, oleophobic, resistant to organic solvents and tested for biosafety and low cytotoxicity.
  • Independent 3rd party tests validate protection against circulating microorganisms (when used as directed)
Each Cell Locker Chamber holds 9 each T-75 cell culture flasks, 20 each 6-well plates, or 24 each 96-well plates.
  • Dishes and flasks can be removed individually on a work tray or inside the entire Cell Locker Chamber with optional transport cover, thus protecting samples from outside air.
  • The non skid tray slides out to allow access to culture vessels, or the entire tray can be removed.
  • The Cell Locker Chamber can be manually disinfected or can be autoclaved.
Warranty: 1 year
TRUSTED_SUSTAINABILITY
Specifications
Description Cell Locker Chamber with copper tray
Chamber Material Stainless Steel
Holds 9 each T-75 cell culture flasks, 20 each 6-well plates, or 24 each 96- well plates
Lighting LED
Material Stainless Steel
No. of Places 53
No. of Shelves 1
Type Cell Locker Chamber
Includes Copper Tray
Warranty 1 Year