Learn More
Abcam Cell Lysis Buffer, 15ML
Catalog No. 502583051
Encompass_Preferred
ab152163 is a 10X Cell Lysis Buffer used to lyse cells under non-denaturing conditions.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3051 Supplier Abcam Supplier No. AB15216315ML
ab152163 is a 10X Cell Lysis Buffer used to lyse cells under non-denaturing conditions.
The product is subject to the following: Abcam Restricted Use Statement