Learn More
Cell Biolabs Inc CELL MIGRATION ASSAY
Catalog No. NC9616141
Encompass_Preferred
The CytoSelect 24-Well Cell Migration Assay Kit contains twelve 8 um polycarbonate membrane inserts in a 24-well plate, which migratory cells are able to pass through. Quantify cells using the provided cell stain solution.
Catalog No. NC9616141 Supplier Cell Biolabs Inc Supplier No. CBA100
Add to Cart
Add to Cart
The CytoSelect 24-Well Cell Migration Assay Kit contains twelve 8 um polycarbonate membrane inserts in a 24-well plate, which migratory cells are able to pass through. Quantify cells using the provided cell stain solution.