Learn More
Bachem H-Cys(NPys)-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-NH2
SDP
Supplier:  Bachem 4091599.0001
Encompass_Preferred
H-Cys(NPys)-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-NH2 Trifluoroacetate, 1417221-06-7, C62H118N40O12S2, Mr 1680.01, 1mg. Cys(Npys)-(Arg)9. Cell-permeable nonaarginine amide, which can be easily conjugated to a cargo molecule, e.g. by maleimide-thiol coupling.
Catalog No. 50-194-6813