Learn More
Eurofins Discoverx AssayComplete Cell Plating 13 Reagent, 100. (93-0563R13A)
Catalog No. 502000802
Encompass
AssayComplete Cell Plating 13 Reagent is an optimized cell plating reagent for use in plating of Eurofins DiscoverX cell lines for assays. This reagent was optimized for maximal cell health and ligand responsiveness during assays while eliminating hormones and growth components which may lead to increased basal activity. This product is provided as a bottle of 100 mL. Please refer to cell line specific datasheet to determine the recommended cell plating reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. 50-200-0802 Supplier Eurofins Discoverx Supplier No. 930563R13A
AssayComplete Cell Plating 13 Reagent is an optimized cell plating reagent for use in plating of Eurofins DiscoverX cell lines for assays. This reagent was optimized for maximal cell health and ligand responsiveness during assays while eliminating hormones and growth components which may lead to increased basal activity. This product is provided as a bottle of 100 mL. Please refer to cell line specific datasheet to determine the recommended cell plating reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.