Learn More
New England Biolabs, Inc. Luna Cell Ready Probe One-Step RT-qPCR Kit – 100 reactions
SDP
Catalog No. NC1773543
Encompass
The Luna Cell Ready Probe One-Step RT-qPCR Kit provides all the necessary components for direct, probe-based RNA detection and quantitation, bypassing the need for RNA extraction and purification. The Luna Cell Ready Probe One-Step RT-qPCR Kit is composed of: 1) the Luna Cell Ready Lysis Module and 2) the Luna Universal Probe One-Step RT-qPCR Kit. Coordinating the actions of DNase I and the Luna Cell Ready Protease, the Luna Cell Ready Lysis Module offers a simple alternative workflow resulting in effective cell lysis, RNA release, and genomic DNA removal simultaneously in a 15-minute protocol. The Lysis Module includes a unique Luna Cell Ready RNA Protection Reagent that maintains RNA integrity during cell lysis. The lysis capacity spans 10 -100,000 cells in a 50 µl lysis reaction. Up to 2 µl of lysate (equivalent to RNA from 0.2 - 4,000 cells) can be transferred into 20 µl downstream RT-qPCR reactions.
Catalog No. NC1773543 Supplier New England Biolabs, Inc. Supplier No. E3031S
The Luna Cell Ready Probe One-Step RT-qPCR Kit provides all the necessary components for direct, probe-based RNA detection and quantitation, bypassing the need for RNA extraction and purification. The Luna Cell Ready Probe One-Step RT-qPCR Kit is composed of: 1) the Luna Cell Ready Lysis Module and 2) the Luna Universal Probe One-Step RT-qPCR Kit. Coordinating the actions of DNase I and the Luna Cell Ready Protease, the Luna Cell Ready Lysis Module offers a simple alternative workflow resulting in effective cell lysis, RNA release, and genomic DNA removal simultaneously in a 15-minute protocol. The Lysis Module includes a unique Luna Cell Ready RNA Protection Reagent that maintains RNA integrity during cell lysis. The lysis capacity spans 10 -100,000 cells in a 50 µl lysis reaction. Up to 2 µl of lysate (equivalent to RNA from 0.2 - 4,000 cells) can be transferred into 20 µl downstream RT-qPCR reactions.