Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Cell Recovery Solution
GREENER_CHOICE

Catalog No. 08774405 Shop All Corning Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-774-405 500 mL
CB-40253 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-774-405 Supplier Corning™ Supplier No. 354270
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Allows for the recovery of cells cultured on Corning™ Matrigel™ Basement Membrane Matrix for subsequent biochemical analyses

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Product Type Cell Recovery Solution
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.