Learn More
Biotang Inc Sarstedt* Cell scraper with soft, flexible bladeL:16cm B:13mm, for T-25, T-75 or dishes 2-pos 100/cs 1/PK single packed Sterile.
Catalog No. 50148370 Shop All BIOTANG Inc Products
Encompass_Preferred
Sarstedt* Cell scraper with soft, flexible bladeL:16cm B:13mm, for T-25, T-75 or dishes 2-pos 100/cs 1/PK single packed Sterile.
Catalog No. 50-148-370 Supplier Biotang Inc Supplier No. 831832
Sarstedt* Cell scraper with soft, flexible bladeL:16cm B:13mm, for T-25, T-75 or dishes 2-pos 100/cs 1/PK single packed Sterile.