Learn More
Chemglass Life Sciences Cell Scraper, 220mm Handle, 13mm Blade, Individually Wrapped, Sterile
SDP
Catalog No. 501897691
Encompass_Preferred
Cell scrapers are for tissue culture harvesting from flasks, plates, or dishes in western or enzyme assays. Designed to gently remove cells, scrapers maximize sample recovery. Improved designs to the blade and handle provide maximum accessibility to the growth surface in a variety of culture vessels.• Improved designs to the blade and handle provide maximum accessibility in a variety of culture vessels • Two sizes available • Easy-tear sterile packing • Sterilized by E-Beam, non-pyrogenic and DNase/RNase-free
Catalog No. 50-189-7691 Supplier Chemglass Life Sciences Supplier No. CGN2403013
Cell scrapers are for tissue culture harvesting from flasks, plates, or dishes in western or enzyme assays. Designed to gently remove cells, scrapers maximize sample recovery. Improved designs to the blade and handle provide maximum accessibility to the growth surface in a variety of culture vessels.• Improved designs to the blade and handle provide maximum accessibility in a variety of culture vessels • Two sizes available • Easy-tear sterile packing • Sterilized by E-Beam, non-pyrogenic and DNase/RNase-free