Learn More
CELLTREAT Scientific Products Cell Scraper, 39cm Handle, 30mm Scraper Blade, Sterile
SDP
Catalog No. 50201976
Encompass_Preferred
CELLTREAT Scientific Products 39cm Cell Scrapers. Cell Scrapers are designed to gently harvest cells from tissue culture flasks, dishes, or bottles. Soft, pliable blade won't damage cells or scratch surfaces. Scraper blade is positioned parallel to the handle for windshield wiper like scraping. Handle length 39cm and blade length 30mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped, 100/case.
Catalog No. 50-201-976 Supplier CELLTREAT Scientific Products Supplier No. 229315
CELLTREAT Scientific Products 39cm Cell Scrapers. Cell Scrapers are designed to gently harvest cells from tissue culture flasks, dishes, or bottles. Soft, pliable blade won't damage cells or scratch surfaces. Scraper blade is positioned parallel to the handle for windshield wiper like scraping. Handle length 39cm and blade length 30mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped, 100/case.