Learn More
Sarstedt Inc Cell scraper, 2-position blade, size: L
Catalog No. NC1890484 Shop All Sarstedt Inc. Products
Encompass_Preferred
Cell scraper, 2-position blade, size: L, handle length: 36 cm, blade length: 1.7 cm, sterile, non-pyrogenic/endotoxin-free, non-cytotoxic, 1 unit/blister
This item is not returnable. View return policy
Catalog No. NC1890484 Supplier Sarstedt Inc Supplier No. 83.3952
Cell scraper, 2-position blade, size: L, handle length: 36 cm, blade length: 1.7 cm, sterile, non-pyrogenic/endotoxin-free, non-cytotoxic, 1 unit/blister