Learn More
Bellco Glass, Inc. Cell Scraper Lifter 25cm 20mm
SDP
Catalog No. 502329980
Encompass_Preferred
Cell Scrapers are designed to gently harvest cells from tissue culture flasks dishes or bottles. Soft pliable blade won’t damage cells or scratch surfaces. Scraper blade is positioned parallel to the handle for windshield wiper like scraping. Lifter blade is positioned perpendicular to the handle for use with a raking motion. Pivoting blade Cell Scraper features free rotating blade that adjusts to the desired position providing access to all corners of the flask or tissue culture vessel.Specifications Handle Length 25cm Blade Length 20mm Blade Orientation Lifter Perpendicular Case Quantity 100
Catalog No. 50-232-9980 Supplier Bellco Glass, Inc. Supplier No. 229311000
Cell Scrapers are designed to gently harvest cells from tissue culture flasks dishes or bottles. Soft pliable blade won’t damage cells or scratch surfaces. Scraper blade is positioned parallel to the handle for windshield wiper like scraping. Lifter blade is positioned perpendicular to the handle for use with a raking motion. Pivoting blade Cell Scraper features free rotating blade that adjusts to the desired position providing access to all corners of the flask or tissue culture vessel.Specifications Handle Length 25cm Blade Length 20mm Blade Orientation Lifter Perpendicular Case Quantity 100