Learn More
STEMCELL Technologies Cell Scraper, Pivoting Blade, 30 cm, Size: 100 Units
SDP
Catalog No. 502390256
Encompass
Use Cell Scraper Pivoting Blade 30 cm for harvesting cells or cell lysates. This sterile RNase/DNase-free and non-pyrogenic blade comes individually wrapped and is best used with Tissue Culture-Treated Dishes (Catalog 38046) and Cell Culture Flasks (Catalog 200-0500/200-0501). It has a sturdy acrylonitrile butadiene styrene (ABS) handle and a flexible thermoplastic elastomer (TPE) blade that prevents damaging cells or scratching surfaces. The pivoting blade rotates to the desired position providing access to all corners of the flask or tissue culture vessel.
Catalog No. 50-239-0256 Supplier STEMCELL Technologies Supplier No. 2000598
Use Cell Scraper Pivoting Blade 30 cm for harvesting cells or cell lysates. This sterile RNase/DNase-free and non-pyrogenic blade comes individually wrapped and is best used with Tissue Culture-Treated Dishes (Catalog 38046) and Cell Culture Flasks (Catalog 200-0500/200-0501). It has a sturdy acrylonitrile butadiene styrene (ABS) handle and a flexible thermoplastic elastomer (TPE) blade that prevents damaging cells or scratching surfaces. The pivoting blade rotates to the desired position providing access to all corners of the flask or tissue culture vessel.