Learn More
Chemglass Life Sciences Cell Screen, 100 Mesh, SST
SDP
Catalog No. 501532420
Encompass_Preferred
Cell Screen, 100 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 100 mesh is equivalent to 149 micron and 0.0059 inches.
Catalog No. 50-153-2420 Supplier Chemglass Life Sciences Supplier No. CLS5023100
Cell Screen, 100 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 100 mesh is equivalent to 149 micron and 0.0059 inches.