Learn More
Chemglass Life Sciences Cell Screen, 150 Mesh, SST
SDP
Catalog No. 501532421
Encompass_Preferred
Cell Screen, 150 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 150 mesh is equivalent to 105 micron and 0.0041 inches.
Catalog No. 50-153-2421 Supplier Chemglass Life Sciences Supplier No. CLS5023150
Add to Cart
Add to Cart
Cell Screen, 150 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 150 mesh is equivalent to 105 micron and 0.0041 inches.