Learn More
Chemglass Life Sciences Cell Screen, 20 Mesh, SST
SDP
Catalog No. 501532414
Encompass_Preferred
Cell Screen, 20 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 20 mesh is equivalent to 841 micron and 0.0331 inches.
Catalog No. 50-153-2414 Supplier Chemglass Life Sciences Supplier No. CLS5023020
Cell Screen, 20 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 20 mesh is equivalent to 841 micron and 0.0331 inches.