Learn More
Chemglass Life Sciences Cell Screen, 200 Mesh, SST
SDP
Catalog No. 501532422
Encompass_Preferred
Cell Screen, 200 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 200 mesh is equivalent to 74 micron and 0.0029 inches.
Catalog No. 50-153-2422 Supplier Chemglass Life Sciences Supplier No. CLS5023200
Cell Screen, 200 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 200 mesh is equivalent to 74 micron and 0.0029 inches.