Learn More
Chemglass Life Sciences Cell Screen, 40 Mesh, SST
SDP
Catalog No. 501532416
Encompass_Preferred
Cell Screen, 40 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 40 mesh is equivalent to 420 micron and 0.0165 inches.
Catalog No. 50-153-2416 Supplier Chemglass Life Sciences Supplier No. CLS5023040
Cell Screen, 40 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 40 mesh is equivalent to 420 micron and 0.0165 inches.