Learn More
Chemglass Life Sciences Cell Screen, 500 Mesh, SST
SDP
Catalog No. 501532425
Encompass_Preferred
Cell Screen, 500 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 500 mesh is equivalent to 25 micron and 0.0010 inches.
Catalog No. 50-153-2425 Supplier Chemglass Life Sciences Supplier No. CLS5023500
Cell Screen, 500 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 500 mesh is equivalent to 25 micron and 0.0010 inches.