Learn More
Bellco Glass, Inc. Cell Screen/50mesh 280
SDP
Catalog No. 501945213
Encompass_Preferred
Prepare cell suspensions and gel extraction of proteins. The stainless steel screens are held in place by a retaining ring on the bottom of the pan that is tightened using the screen replacement key. The screens are available in different mesh sizes and have a diameter of 37mm. MADE IN THE USA. Specifications:Stainless Steel Size: 280Case Quantity: 5
Catalog No. 50-194-5213 Supplier Bellco Glass, Inc. Supplier No. 198500050
Prepare cell suspensions and gel extraction of proteins. The stainless steel screens are held in place by a retaining ring on the bottom of the pan that is tightened using the screen replacement key. The screens are available in different mesh sizes and have a diameter of 37mm. MADE IN THE USA. Specifications:Stainless Steel Size: 280Case Quantity: 5