Learn More
CELLTREAT Scientific Products Cell Scraper, 25cm Handle, 12mm Narrow Blade, Sterile
SDP
Catalog No. NC3377713
Encompass_Preferred
Cell Scrapers are designed to gently harvest cells from tissue culture flasks, dishes, or bottlesSoft, pliable blade won’t damage cells or scratch surfaces; outperforms hard plastic blade of other brandsScraper blade is positioned parallel to the handle for windshield wiper like scrapingLifter blade is positioned perpendicular to the handle for use with a raking motionPivoting Blade Cell Scraper features free rotating blade that adjusts to the desired position, providing access to all corners of the flask or tissue culture vessel (PN 229312)12mm Narrow Blade is ideal for accessing cell culture vessels with smaller openings, such as 12.5cm2, 25cm2, and 75cm2 Tissue Culture Flasks
Catalog No. NC3377713 Supplier CELLTREAT Scientific Products Supplier No. 229317
Cell Scrapers are designed to gently harvest cells from tissue culture flasks, dishes, or bottlesSoft, pliable blade won’t damage cells or scratch surfaces; outperforms hard plastic blade of other brandsScraper blade is positioned parallel to the handle for windshield wiper like scrapingLifter blade is positioned perpendicular to the handle for use with a raking motionPivoting Blade Cell Scraper features free rotating blade that adjusts to the desired position, providing access to all corners of the flask or tissue culture vessel (PN 229312)12mm Narrow Blade is ideal for accessing cell culture vessels with smaller openings, such as 12.5cm2, 25cm2, and 75cm2 Tissue Culture Flasks