Learn More
Chemglass Life Sciences Cell Separator Stand, Magnetic, 8.5 x 3", Holds (24) 1.5 or 2.0mL Tubes
SDP
Catalog No. 501311354
Encompass
Cell Separator Stand, Magnetic, 8.5 x 3", Holds (24) 1.5 or 2.0mL Tubes. Magnetic Stand for DNA and RNA extraction or cell purification. This magnetic stand is designed to be used with 96 well multi well plates for magnetic bead based cell purification or DNA and RNA extraction. Cells or nucleic acid tagged with paramagnetic beads will be held down on the bottom of the plate by powerful magnetic force for easy removal of media or reagents for cell purification or nucleic acid extraction procedure. Made of clear acrylic.
Catalog No. 50-131-1354 Supplier Chemglass Life Sciences Supplier No. CLS4284024
Cell Separator Stand, Magnetic, 8.5 x 3", Holds (24) 1.5 or 2.0mL Tubes. Magnetic Stand for DNA and RNA extraction or cell purification. This magnetic stand is designed to be used with 96 well multi well plates for magnetic bead based cell purification or DNA and RNA extraction. Cells or nucleic acid tagged with paramagnetic beads will be held down on the bottom of the plate by powerful magnetic force for easy removal of media or reagents for cell purification or nucleic acid extraction procedure. Made of clear acrylic.