Learn More
STEMCELL Technologies EasySep™ Human T Cell Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC9959219
Encompass
The EasySep™ Human T Cell Enrichment Kit is designed to isolate T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-T cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. For even faster cell isolations, we recommend the new EasySep™ Human T Cell Isolation Kit (17951) (http://www.stemcell.com/en/Products/All-Products/EasySep-Human-T-Cell-Isolation-Kit.aspx) which isolates cells in just 8 minutes.
Catalog No. NC9959219 Supplier STEMCELL Technologies Supplier No. 19051
The EasySep™ Human T Cell Enrichment Kit is designed to isolate T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-T cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. For even faster cell isolations, we recommend the new EasySep™ Human T Cell Isolation Kit (17951) (http://www.stemcell.com/en/Products/All-Products/EasySep-Human-T-Cell-Isolation-Kit.aspx) which isolates cells in just 8 minutes.