Learn More
MilliporeSigma MILLIPLEX MAP TGFbeta Signaling Pathway Magnetic Bead 6-Plex - Cell Signaling Multiplex Assay
Catalog No. NC1079330
Encompass
Intracellular Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous relative quantitation of multiple phosphorylation and total pathway proteins in tissue and cell lysate samples. Compare Multiplexing results to those of Western blotting.
Catalog No. NC1079330 Supplier MilliporeSigma Supplier No. 48614MAG
Intracellular Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous relative quantitation of multiple phosphorylation and total pathway proteins in tissue and cell lysate samples. Compare Multiplexing results to those of Western blotting.