Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Full Moon Biosystems Cell Signaling Phospho Antibody Array, ELISA-based, 304 antibodies, 2 array slides, 2 samples, 16 pathways, 6 antibody replicates
SDP

Supplier:  Full Moon Biosystems PCS300

Encompass

The Full Moon BioSystems PCS300 Cell Signaling Phospho Antibody Array is a high-throughput platform designed for comprehensive protein phosphorylation profiling in scientific research. This ELISA-based array enables qualitative profiling and screening of candidate biomarkers across 16 critical cell signaling pathways, facilitating the identification of total protein and phosphorylation changes between normal and treated/diseased samples. Features include:

  • 304 site-specific and phospho-specific antibodies
  • 6 replicates per antibody
  • Human reactivity: 99%, Mouse: 87%, Rat: 66%
  • Detection method optimized for fluorescence
  • 3D polymer-coated glass slide for solid support
  • Each package contains 2 array slides
  • Suitable for cell and tissue lysates
  • Internal controls include Beta-actin and GAPDH
  • Slide dimensions: 76 x 25 x 1 mm
  • Storage condition: 4°C for 6 months

Catalog No. NC2126421


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.