Learn More
YMC America, Inc Cell-SJ guard 3um 10x4mm
SDP
Catalog No. 5000879510
Encompass
YMC s CHIRAL ART series features polysaccharide-derived chiral stationary phases (CSPs) based on amylose and cellulose tris-carbamate derivatives available in both coated and immobilized formats These columns provide exceptional stereoselectivity durability and solvent versatility supporting normal-phase reversed-phase and supercritical fluid chromatography (SFC) modes
Catalog No. 50-008-79510 Supplier YMC America, Inc Supplier No. KSJ99S030104GC
YMC s CHIRAL ART series features polysaccharide-derived chiral stationary phases (CSPs) based on amylose and cellulose tris-carbamate derivatives available in both coated and immobilized formats These columns provide exceptional stereoselectivity durability and solvent versatility supporting normal-phase reversed-phase and supercritical fluid chromatography (SFC) modes