Learn More
Perkin Elmer US LLC Cell Spacer - 1mm
Catalog No. 509045911
Encompass_Preferred
Cell Spacers of varying thicknesses allow cells with 1 mm, 2 mm, or 5 mm optical path lengths to fit securely into a 10 mm cell holder.
Catalog No. 50-904-5911 Supplier Perkin Elmer US LLC Supplier No. N9302741
Cell Spacers of varying thicknesses allow cells with 1 mm, 2 mm, or 5 mm optical path lengths to fit securely into a 10 mm cell holder.