Learn More
Research Products International Corp Cell Spreaders L Shaped S, 10 per Package 500/cs
SDP
Catalog No. 502230560
Encompass
These cell spreaders have an L shape that provides quick and uniform cell distribution across surface of growth media. Spreaders are made from durable polystyrene for easy disposal after each use.
Supplied in sealed pouches, sterile.
Catalog No. 50-223-0560 Supplier Research Products International Corp Supplier No. 247660A
These cell spreaders have an L shape that provides quick and uniform cell distribution across surface of growth media. Spreaders are made from durable polystyrene for easy disposal after each use.
Supplied in sealed pouches, sterile.