Learn More
Thomas Scientific CELL SPREADER0
SDP
Catalog No. NC9098345
Encompass
Disposable, high-impact polystyrene No flame sterilization needed “T” design for easy manipulation and spreading of bacteria onto the surface of culture plates. Applies the appropriate volume (0.05-0.1 mL ) of the bacterial sample onto the plate. Supplied sterile in individual packs. Total length of handle is 171 mm and width of spreader is 36 mm.
Catalog No. NC9098345 Supplier Thomas Scientific Supplier No. 7012Q40
Disposable, high-impact polystyrene No flame sterilization needed “T” design for easy manipulation and spreading of bacteria onto the surface of culture plates. Applies the appropriate volume (0.05-0.1 mL ) of the bacterial sample onto the plate. Supplied sterile in individual packs. Total length of handle is 171 mm and width of spreader is 36 mm.