Learn More
Chemglass Life Sciences Cell Spreader, L-Shaped, Orange, Sterile
SDP
Catalog No. 501897544
Encompass
• L-shape spreaders made of polystyrene• Smooth surface and angled end reduce risk of damaging culture medium• L-shape allows application on the entire culture dish or plate surface• 10 per bag (sterile, resealable)• Gamma radiation sterilized
Catalog No. 50-189-7544 Supplier Chemglass Life Sciences Supplier No. CGE1351001
• L-shape spreaders made of polystyrene• Smooth surface and angled end reduce risk of damaging culture medium• L-shape allows application on the entire culture dish or plate surface• 10 per bag (sterile, resealable)• Gamma radiation sterilized