Learn More
Research Products International Corp Cell Spreader, T Shaped, Sterile, Individually Wrapped, 100 per Case
SDP
Catalog No. 50476531
Encompass
These cell spreaders have an T shape that provides quick and uniform cell distribution across surface of growth media. Spreaders are made from durable polystyrene for easy disposal after each use.
Supplied in sealed pouches, sterile.
Catalog No. 50-476-531 Supplier Research Products International Corp Supplier No. 247663
These cell spreaders have an T shape that provides quick and uniform cell distribution across surface of growth media. Spreaders are made from durable polystyrene for easy disposal after each use.
Supplied in sealed pouches, sterile.